Product Details

SNP ID
rs201415317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126271607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCCTATTGGCTGAAGAAGCTG[A/G]AGAGCAGACGAGGTGCTATTCGAGT
Phenotype
MIM: 616587 MIM: 613622 MIM: 182180 MIM: 606252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM118B PubMed Links

Gene Details

Gene
FAM118B
Gene Name
family with sequence similarity 118 member B
There are no transcripts associated with this gene.

Gene
FOXRED1
Gene Name
FAD dependent oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017547.3 1555 Missense Mutation AAG,GAG K86E NP_060017.1
XM_006718879.3 1555 Intron XP_006718942.1
XM_006718881.3 1555 Intron XP_006718944.1
XM_011542895.2 1555 UTR 5 XP_011541197.1
XM_011542896.2 1555 UTR 5 XP_011541198.1
XM_017018000.1 1555 Missense Mutation AAG,GAG K86E XP_016873489.1
XM_017018001.1 1555 UTR 5 XP_016873490.1
XM_017018002.1 1555 Intron XP_016873491.1
XM_017018003.1 1555 UTR 5 XP_016873492.1
XM_017018004.1 1555 UTR 5 XP_016873493.1
XM_017018005.1 1555 UTR 5 XP_016873494.1
XM_017018006.1 1555 UTR 5 XP_016873495.1
Gene
SRPRA
Gene Name
SRP receptor alpha subunit
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
There are no transcripts associated with this gene.

View Full Product Details