Product Details

SNP ID
rs201237944
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:13492523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTTCGGGGCCTCTGGGAACCA[G/T]CATCTCTGGGAGCTAGAGGAGCTCC
Phenotype
MIM: 168450
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTH PubMed Links

Gene Details

Gene
PTH
Gene Name
parathyroid hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000315.3 369 Missense Mutation GAT,GCT D77A NP_000306.1
NM_001316352.1 369 Missense Mutation GAT,GCT D109A NP_001303281.1

View Full Product Details