Product Details

SNP ID
rs201424323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56462498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTCACATATGTCTTGGTGATTG[C/T]TTTGTTGACTGCTTCTTTTACATCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5M3 PubMed Links

Gene Details

Gene
OR5M3
Gene Name
olfactory receptor family 5 subfamily M member 3
There are no transcripts associated with this gene.

Gene
OR5M9
Gene Name
olfactory receptor family 5 subfamily M member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004743.1 904 Missense Mutation ACA,GCA T302A NP_001004743.1

View Full Product Details