Product Details

SNP ID
rs201172638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67306874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCACCCCGGCCTCCCCGGCTC[C/T]GCTACCTGCTGGTAGTTTCTACACG
Phenotype
MIM: 615126 MIM: 606780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13D PubMed Links

Gene Details

Gene
ANKRD13D
Gene Name
ankyrin repeat domain 13D
There are no transcripts associated with this gene.

Gene
SSH3
Gene Name
slingshot protein phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017857.3 554 Missense Mutation CGC,TGC R126C NP_060327.3
XM_017017942.1 554 Missense Mutation CGC,TGC R26C XP_016873431.1

View Full Product Details