Product Details

SNP ID
rs199475696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102840453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGGGTATTGTCCAAGACCTCA[G/A]TCCTTTGGGTGTATGGGTCGTAGCG
Phenotype
MIM: 612349
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PAH PubMed Links

Gene Details

Gene
PAH
Gene Name
phenylalanine hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000277.1 1734 Missense Mutation ACT,ATT T421I NP_000268.1
XM_017019370.1 1734 Intron XP_016874859.1

View Full Product Details