Product Details

SNP ID
rs199578843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8649514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCATCTTACCTTTCATAGCTTC[A/G]TGTTCCTTACAGACAAGACGAGAGC
Phenotype
MIM: 601103
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFAP5 PubMed Links

Gene Details

Gene
MFAP5
Gene Name
microfibrillar associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297709.1 630 Silent Mutation CAC,CAT H122H NP_001284638.1
NM_001297710.1 630 Silent Mutation CAC,CAT H110H NP_001284639.1
NM_001297711.1 630 Silent Mutation CAC,CAT H107H NP_001284640.1
NM_001297712.1 630 Intron NP_001284641.1
NM_003480.3 630 Silent Mutation CAC,CAT H132H NP_003471.1

View Full Product Details