Product Details

SNP ID
rs201009829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6770189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGAAGAAGAGGATGATGAAGGG[C/G]CCGCGCTGAAGAGAGCTGCCGAAGA
Phenotype
MIM: 153337 MIM: 168440
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LAG3 PubMed Links

Gene Details

Gene
LAG3
Gene Name
lymphocyte activating 3
There are no transcripts associated with this gene.

Gene
PTMS
Gene Name
parathymosin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011520987.1 873 Silent Mutation GGC,GGG G102G XP_011519289.1

View Full Product Details