Product Details

SNP ID
rs200350630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8943415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGCATACTCACGTGACAAGTA[A/C]GATGGAACCCACACTGAGGTGGGAG
Phenotype
MIM: 154540 MIM: 602978
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
M6PR PubMed Links

Gene Details

Gene
M6PR
Gene Name
mannose-6-phosphate receptor, cation dependent
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207024.1 849 Intron NP_001193953.1
NM_002355.3 849 Missense Mutation GTA,TTA V192L NP_002346.1
XM_005253376.2 849 Missense Mutation GTA,TTA V192L XP_005253433.1
Gene
PHC1
Gene Name
polyhomeotic homolog 1
There are no transcripts associated with this gene.

View Full Product Details