Product Details

SNP ID
rs200357192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:565972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAATGACCACGAGCAGGACACC[A/G]ATGACCACCTGCAGGAGCAGAGAGA
Phenotype
MIM: 612220 MIM: 607297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALNT3 PubMed Links

Gene Details

Gene
B4GALNT3
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
There are no transcripts associated with this gene.

Gene
NINJ2
Gene Name
ninjurin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294345.1 621 Silent Mutation ATC,ATT I73I NP_001281274.1
NM_001294346.1 621 Silent Mutation ATC,ATT I44I NP_001281275.1
NM_016533.5 621 Silent Mutation ATC,ATT I126I NP_057617.2

View Full Product Details