Product Details

SNP ID
rs200165002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:92706746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGGAGTCTCCTCCAGTTTGAC[C/T]GGCAAGCCCCAGGCCGCATCTCCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf74 PubMed Links

Gene Details

Gene
C12orf74
Gene Name
chromosome 12 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037671.3 566 Missense Mutation CGG,TGG R39W NP_001032760.1
NM_001178097.2 566 Missense Mutation CGG,TGG R39W NP_001171568.1

View Full Product Details