Product Details
- SNP ID
-
rs201607310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52822059 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGCGCCACCTCCGCACACAGTG[A/G]TGGAGTTGCCAGTCACAGCTGCAAA
- Phenotype
-
MIM: 123940
MIM: 611160
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT4
PubMed Links
Gene Details
- Gene
- KRT4
- Gene Name
- keratin 4
There are no transcripts associated with this gene.
- Gene
- KRT79
- Gene Name
- keratin 79
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175834.2 |
1454 |
Missense Mutation |
ACC,ATC |
T474I |
NP_787028.1 |
View Full Product Details