Product Details

SNP ID
rs201681638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7715290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATCGGAAGCTTTACTCCGTCGA[C/G]AGTCTGTAGGAGCAGCAGTCCTCAG
Phenotype
MIM: 608408
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DPPA3 PubMed Links

Gene Details

Gene
DPPA3
Gene Name
developmental pluripotency associated 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199286.3 Intron NP_954980.1

View Full Product Details