Product Details

SNP ID
rs201715919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:66302684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATCTGGTGGAGGAGGACGACGA[A/C]TACCTAAACGACGACGTGGAGGAGG
Phenotype
MIM: 614539
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HELB PubMed Links

Gene Details

Gene
HELB
Gene Name
DNA helicase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033647.4 140 Missense Mutation GAA,GAC E27D NP_387467.2
XM_005269234.2 140 Missense Mutation GAA,GAC E27D XP_005269291.1
XM_017020230.1 140 Missense Mutation GAA,GAC E27D XP_016875719.1

View Full Product Details