Product Details

SNP ID
rs201598330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56923941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAGTTTGGCATCCAGGCCAGG[G/T]TTGTAGCGGATGCGAGGGCTCCGGG
Phenotype
MIM: 609769
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SDR9C7 PubMed Links

Gene Details

Gene
SDR9C7
Gene Name
short chain dehydrogenase/reductase family 9C, member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_148897.2 978 Silent Mutation NP_683695.1

View Full Product Details