Product Details

SNP ID
rs202194762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49792480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGCAGCCCCTCCAGTTGTCGG[G/T]GTGGGCACATGAGAGGGTCCGAGGA
Phenotype
MIM: 615104
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NCKAP5L PubMed Links

Gene Details

Gene
NCKAP5L
Gene Name
NCK associated protein 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037806.3 4255 Missense Mutation CAC,CCC H1253P NP_001032895.2
XM_005269050.3 4255 Missense Mutation CAC,CCC H1253P XP_005269107.1
XM_006719525.2 4255 Missense Mutation CAC,CCC H1253P XP_006719588.1
XM_011538610.2 4255 Missense Mutation CAC,CCC H1253P XP_011536912.1
XM_011538611.2 4255 Missense Mutation CAC,CCC H1253P XP_011536913.1

View Full Product Details