Product Details
- SNP ID
-
rs201279046
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:131961895 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTAGGCAGAATGATTTGGACATT[A/G]AAGAAGAGGAGGAGGAGGAGGAAGA
- Phenotype
-
MIM: 606265
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EP400
PubMed Links
Gene Details
- Gene
- EP400
- Gene Name
- E1A binding protein p400
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015409.4 |
1428 |
Missense Mutation |
AAA,GAA |
K426E |
NP_056224.3 |
View Full Product Details