Product Details

SNP ID
rs201262620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:75481843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTATTTTTGCAGACTTGGGAC[C/T]CAGCACTAGCCCAAATTGCAAAAGC
Phenotype
MIM: 602692
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLIPR1 PubMed Links

Gene Details

Gene
GLIPR1
Gene Name
GLI pathogenesis related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006851.2 311 Missense Mutation CCA,TCA P62S NP_006842.2
XM_011537775.2 311 Missense Mutation CCA,TCA P62S XP_011536077.1
XM_017018721.1 311 UTR 5 XP_016874210.1

View Full Product Details