Product Details
- SNP ID
-
rs201257827
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:40933601 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTACTTCCTGTGTAAGAGTTTTTG[C/T]TTCTTGAGTTTTGATAACTGAAGTG
- Phenotype
-
MIM: 189973
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ELF1
PubMed Links
Gene Details
- Gene
- ELF1
- Gene Name
- E74 like ETS transcription factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145353.1 |
2286 |
Missense Mutation |
ACA,GCA |
T538A |
NP_001138825.1 |
NM_172373.3 |
2286 |
Missense Mutation |
ACA,GCA |
T562A |
NP_758961.1 |
XM_005266276.1 |
2286 |
Missense Mutation |
ACA,GCA |
T562A |
XP_005266333.1 |
XM_005266277.3 |
2286 |
Missense Mutation |
ACA,GCA |
T562A |
XP_005266334.1 |
XM_011534950.1 |
2286 |
Missense Mutation |
ACA,GCA |
T562A |
XP_011533252.1 |
XM_011534951.2 |
2286 |
Missense Mutation |
ACA,GCA |
T503A |
XP_011533253.1 |
XM_017020409.1 |
2286 |
Missense Mutation |
ACA,GCA |
T562A |
XP_016875898.1 |
XM_017020410.1 |
2286 |
Missense Mutation |
ACA,GCA |
T562A |
XP_016875899.1 |
XM_017020411.1 |
2286 |
Missense Mutation |
ACA,GCA |
T562A |
XP_016875900.1 |
XM_017020412.1 |
2286 |
Missense Mutation |
ACA,GCA |
T562A |
XP_016875901.1 |
XM_017020413.1 |
2286 |
Missense Mutation |
ACA,GCA |
T562A |
XP_016875902.1 |
View Full Product Details