Product Details

SNP ID
rs200686308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:96090963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCCCCCTCCTGCACCAGCTCCT[A/G]CACCAACTTCGGGGAGCAGCCCCGC
Phenotype
MIM: 609401
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HS6ST3 PubMed Links

Gene Details

Gene
HS6ST3
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153456.3 330 Missense Mutation TAC,TGC Y34C NP_703157.2
XM_011521076.2 330 Missense Mutation TAC,TGC Y34C XP_011519378.1
XM_017020543.1 330 Missense Mutation TAC,TGC Y34C XP_016876032.1

View Full Product Details