Product Details

SNP ID
rs201791650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49630458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCAGTGGCCACCATGGGTTCTG[A/T]GAATTCCAGAGGTCACAAGGCGGAA
Phenotype
MIM: 609351
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL11 PubMed Links

Gene Details

Gene
ARL11
Gene Name
ADP ribosylation factor like GTPase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138450.5 255 Missense Mutation GAG,GTG E4V NP_612459.1
XM_005266253.3 255 Missense Mutation GAG,GTG E4V XP_005266310.1

View Full Product Details