Product Details

SNP ID
rs200374851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:96987039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGGAGTAGCAGAATGGTTAGCC[G/T]TCGTGCTTTCTGCTTAAGGCAGCTG
Phenotype
MIM: 606922
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00359 PubMed Links

Gene Details

Gene
LINC00359
Gene Name
long intergenic non-protein coding RNA 359
There are no transcripts associated with this gene.

Gene
OXGR1
Gene Name
oxoglutarate receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080818.3 1102 Silent Mutation AGG,CGG R241R NP_543008.3
XM_005254043.3 1102 Silent Mutation AGG,CGG R241R XP_005254100.1
XM_005254044.3 1102 Silent Mutation AGG,CGG R241R XP_005254101.1
XM_005254045.2 1102 Silent Mutation AGG,CGG R241R XP_005254102.1
XM_005254046.2 1102 Silent Mutation AGG,CGG R241R XP_005254103.1
XM_011521077.2 1102 Silent Mutation AGG,CGG R241R XP_011519379.1

View Full Product Details