Product Details

SNP ID
rs199763203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:96091476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCCGCTTCTCCACCGGCTGGA[A/G]CTGCGGGCTGCACGCCGACTGGACG
Phenotype
MIM: 609401
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HS6ST3 PubMed Links

Gene Details

Gene
HS6ST3
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153456.3 843 Missense Mutation AAC,AGC N205S NP_703157.2
XM_011521076.2 843 Missense Mutation AAC,AGC N205S XP_011519378.1
XM_017020543.1 843 Missense Mutation AAC,AGC N205S XP_016876032.1

View Full Product Details