Product Details

SNP ID
rs200512455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46344788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGAAATGGGAAGATGACAGTC[A/G]TATTCTGGCAAAATTCACAAATAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0226L PubMed Links

Gene Details

Gene
KIAA0226L
Gene Name
KIAA0226 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286761.1 1777 Missense Mutation ACG,ATG T610M NP_001273690.1
NM_001286762.1 1777 Nonsense Mutation CGA,TGA R559* NP_001273691.1
NM_001286763.1 1777 Missense Mutation ACG,ATG T543M NP_001273692.1
NM_001286764.1 1777 Missense Mutation ACG,ATG T475M NP_001273693.1
NM_001286765.1 1777 Missense Mutation ACG,ATG T453M NP_001273694.1
NM_001286766.1 1777 Missense Mutation ACG,ATG T395M NP_001273695.1
NM_025113.3 1777 Missense Mutation ACG,ATG T610M NP_079389.2

View Full Product Details