Product Details

SNP ID
rs201842534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:44962453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCCCCCTCCGCGCTGCTTCTGC[C/T]GCCGCCCTTTCCAGTCCTCTCTACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM179B PubMed Links

Gene Details

Gene
FAM179B
Gene Name
family with sequence similarity 179 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308120.1 235 Missense Mutation CCG,CTG P11L NP_001295049.1
NM_015091.3 235 Missense Mutation CCG,CTG P11L NP_055906.2
XM_011536571.1 235 Missense Mutation CCG,CTG P11L XP_011534873.1
XM_017021098.1 235 Missense Mutation CCG,CTG P11L XP_016876587.1
XM_017021099.1 235 Missense Mutation CCG,CTG P11L XP_016876588.1
XM_017021100.1 235 Missense Mutation CCG,CTG P11L XP_016876589.1
Gene
KLHL28
Gene Name
kelch like family member 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308112.1 235 Intron NP_001295041.1
NM_017658.4 235 Intron NP_060128.2
XM_005267770.3 235 Intron XP_005267827.1
XM_006720174.3 235 Intron XP_006720237.1
XM_011536847.2 235 Intron XP_011535149.1
XM_011536849.2 235 Intron XP_011535151.1
XM_017021378.1 235 Intron XP_016876867.1

View Full Product Details