Product Details

SNP ID
rs201842923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20033985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGTATCTGCTGAAGGGCCAGAC[A/G]TAGATAAAGACACACGGAGCAAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4K13 PubMed Links

Gene Details

Gene
OR4K13
Gene Name
olfactory receptor family 4 subfamily K member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004714.1 774 Silent Mutation TAC,TAT Y258Y NP_001004714.1

View Full Product Details