Product Details

SNP ID
rs201435792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20891807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGAGGGCTCAGTGGTTTGCCATC[C/T]AGCACATCAGTCTGAACCCCCCTCG
Phenotype
MIM: 131398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNASE3 PubMed Links

Gene Details

Gene
RNASE3
Gene Name
ribonuclease A family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002935.2 175 Nonsense Mutation CAG,TAG Q41* NP_002926.2

View Full Product Details