Product Details

SNP ID
rs201764769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:37591776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGTCGCCGTCGCCCCACTGTG[A/G]TCCAGAGTCTGGGGGCTGGCGGCGG
Phenotype
MIM: 602294
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXA1 PubMed Links

Gene Details

Gene
FOXA1
Gene Name
forkhead box A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004496.3 1718 Silent Mutation GAC,GAT D336D NP_004487.2
XM_017021246.1 1718 Silent Mutation GAC,GAT D303D XP_016876735.1
Gene
TTC6
Gene Name
tetratricopeptide repeat domain 6
There are no transcripts associated with this gene.

View Full Product Details