Product Details

SNP ID
rs201447722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94128517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCCTGTCTAGGACACTTACCCA[C/T]GGACTGCAGAGTAGCCACCAGGCTC
Phenotype
MIM: 611319
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFI27L2 PubMed Links

Gene Details

Gene
IFI27L2
Gene Name
interferon alpha inducible protein 27 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032036.2 252 Missense Mutation ATG,GTG M66V NP_114425.1

View Full Product Details