Product Details

SNP ID
rs201741583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74663740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGTCAGAGCATGCCAAAGCCCT[C/G]GCAACCCTCGCTGATGGCCAGCTGC
Phenotype
MIM: 615380
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AREL1 PubMed Links

Gene Details

Gene
AREL1
Gene Name
apoptosis resistant E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039479.1 2744 Missense Mutation CAG,GAG Q818E NP_001034568.1
XM_005268244.3 2744 Missense Mutation CAG,GAG Q657E XP_005268301.1
XM_006720344.2 2744 Missense Mutation CAG,GAG Q808E XP_006720407.1
XM_011537415.2 2744 Missense Mutation CAG,GAG Q818E XP_011535717.1
XM_011537416.2 2744 Missense Mutation CAG,GAG Q758E XP_011535718.1
XM_011537417.2 2744 Missense Mutation CAG,GAG Q657E XP_011535719.1

View Full Product Details