Product Details

SNP ID
rs200194221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104927205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCATGCTCCATGCCGCCCCGCC[G/T]CCCGTGGGACAGAGAGGCTGGCACG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLD4 PubMed Links

Gene Details

Gene
PLD4
Gene Name
phospholipase D family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308174.1 240 Missense Mutation CGC,CTC R29L NP_001295103.1
NM_138790.3 240 Missense Mutation CGC,CTC R22L NP_620145.2
XM_011536411.2 240 Missense Mutation CGC,CTC R29L XP_011534713.1
XM_017020965.1 240 Missense Mutation CGC,CTC R22L XP_016876454.1

View Full Product Details