Product Details

SNP ID
rs199765894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103736145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCGTCACTGTTCTGGGCCTCG[C/T]AGTCCCACAGAGCATACGCCACACC
Phenotype
MIM: 606455 MIM: 613504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R13B PubMed Links

Gene Details

Gene
PPP1R13B
Gene Name
protein phosphatase 1 regulatory subunit 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015316.2 3881 Missense Mutation TAC,TGC Y1030C NP_056131.2
XM_005267487.4 3881 Missense Mutation TAC,TGC Y1098C XP_005267544.1
XM_011536593.2 3881 Missense Mutation TAC,TGC Y1133C XP_011534895.1
XM_017021116.1 3881 Missense Mutation TAC,TGC Y1065C XP_016876605.1
XM_017021117.1 3881 Missense Mutation TAC,TGC Y1062C XP_016876606.1
XM_017021118.1 3881 Missense Mutation TAC,TGC Y1062C XP_016876607.1
XM_017021119.1 3881 Missense Mutation TAC,TGC Y813C XP_016876608.1
Gene
ZFYVE21
Gene Name
zinc finger FYVE-type containing 21
There are no transcripts associated with this gene.

View Full Product Details