Product Details

SNP ID
rs200139158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75663210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATCATCCTCAGAGGATGAGGAG[G/T]TCATAAGTCAAGAGTAAGTAATAGC
Phenotype
MIM: 604576 MIM: 612268
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C14orf1 PubMed Links

Gene Details

Gene
C14orf1
Gene Name
chromosome 14 open reading frame 1
There are no transcripts associated with this gene.

Gene
TTLL5
Gene Name
tubulin tyrosine ligase like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015072.4 336 Missense Mutation GTC,TTC V21F NP_055887.3

View Full Product Details