Product Details

SNP ID
rs199498666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69054153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCCAAGTTCTGCCCGTTATTGT[G/T]AGGGTGAGGGGGACGAGGGTGTAAG
Phenotype
MIM: 603812
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DCAF5 PubMed Links

Gene Details

Gene
DCAF5
Gene Name
DDB1 and CUL4 associated factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284206.1 2749 Missense Mutation NP_001271135.1
NM_001284207.1 2749 Missense Mutation NP_001271136.1
NM_001284208.1 2749 Intron NP_001271137.1
NM_003861.2 2749 Missense Mutation NP_003852.1
XM_006720297.2 2749 Missense Mutation XP_006720360.1
XM_006720298.2 2749 Missense Mutation XP_006720361.1
XM_006720299.3 2749 Missense Mutation XP_006720362.1
XM_006720300.2 2749 Missense Mutation XP_006720363.1
XM_011537278.1 2749 Missense Mutation XP_011535580.1
XM_011537279.2 2749 Missense Mutation XP_011535581.1
XM_011537280.2 2749 Missense Mutation XP_011535582.1
XM_017021733.1 2749 Missense Mutation XP_016877222.1
XM_017021734.1 2749 Missense Mutation XP_016877223.1
XM_017021735.1 2749 Missense Mutation XP_016877224.1
XM_017021736.1 2749 Missense Mutation XP_016877225.1
XM_017021737.1 2749 Missense Mutation XP_016877226.1
XM_017021738.1 2749 Missense Mutation XP_016877227.1
XM_017021739.1 2749 Missense Mutation XP_016877228.1

View Full Product Details