Product Details

SNP ID
rs199611345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76790704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGCATTGTAGTGACCTCTGGGC[A/G]GCCACGCTGGGGCAGGAAGTGGGTA
Phenotype
MIM: 609011
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANGEL1 PubMed Links

Gene Details

Gene
ANGEL1
Gene Name
angel homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015305.3 1872 Missense Mutation CGC,TGC R587C NP_056120.2
XM_017021115.1 1872 UTR 3 XP_016876604.1
Gene
LOC100506603
Gene Name
uncharacterized LOC100506603
There are no transcripts associated with this gene.

Gene
VASH1
Gene Name
vasohibin 1
There are no transcripts associated with this gene.

View Full Product Details