Product Details

SNP ID
rs201478784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34752918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGCGTAGACCTGATGGGTGTG[A/G]AGTCAGCTCCTTAACCTCTAAACAA
Phenotype
MIM: 607058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJD2 PubMed Links

Gene Details

Gene
GJD2
Gene Name
gap junction protein delta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020660.2 619 Missense Mutation CCA,TCA P176S NP_065711.1
XM_017022438.1 619 Missense Mutation CCA,TCA P125S XP_016877927.1
Gene
LOC101928174
Gene Name
uncharacterized LOC101928174
There are no transcripts associated with this gene.

View Full Product Details