Product Details

SNP ID
rs201665956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64072979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCACAATTTCCCGTAAGTTGGG[A/G]TTTTCCATTGCAGCTGCCACTCGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM96A PubMed Links

Gene Details

Gene
FAM96A
Gene Name
family with sequence similarity 96 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014812.2 675 UTR 3 NP_001014812.1
NM_001289108.1 675 UTR 3 NP_001276037.1
NM_032231.5 675 Silent Mutation AAC,AAT N145N NP_115607.1

View Full Product Details