Product Details

SNP ID
rs201154324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50242954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGTGGCCGGGATGAGGAAGAGA[C/T]GGCCAGCTTTAGCTATTTCCTTTAA
Phenotype
MIM: 142704 MIM: 603247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HDC PubMed Links

Gene Details

Gene
HDC
Gene Name
histidine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306146.1 1506 Missense Mutation CAT,CGT H399R NP_001293075.1
NM_002112.3 1506 Missense Mutation CAT,CGT H432R NP_002103.2
XM_017022094.1 1506 Missense Mutation CAT,CGT H467R XP_016877583.1
XM_017022095.1 1506 Missense Mutation CAT,CGT H434R XP_016877584.1
XM_017022096.1 1506 Missense Mutation CAT,CGT H391R XP_016877585.1
XM_017022097.1 1506 Missense Mutation CAT,CGT H388R XP_016877586.1
XM_017022098.1 1506 Missense Mutation CAT,CGT H323R XP_016877587.1
XM_017022099.1 1506 Intron XP_016877588.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
There are no transcripts associated with this gene.

View Full Product Details