Product Details

SNP ID
rs202225656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45366492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAATTTGTGTCTGTCTTCTACA[C/G]AGTGGATGGGATAATCCTAATTGGA
Phenotype
MIM: 602360
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GATM PubMed Links

Gene Details

Gene
GATM
Gene Name
glycine amidinotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321015.1 1021 Missense Mutation TCT,TGT S102C NP_001307944.1
NM_001482.2 1021 Missense Mutation TCT,TGT S231C NP_001473.1

View Full Product Details