Product Details

SNP ID
rs202156060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78092630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGCCTGGGGAGAGATCCTGAG[C/G]GGGCAGGGGACAGCCAGGCCCTGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SH2D7 PubMed Links

Gene Details

Gene
SH2D7
Gene Name
SH2 domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101404.1 46 Missense Mutation CGG,GGG R16G NP_001094874.1
XM_011521895.2 46 Missense Mutation CGG,GGG R16G XP_011520197.1
XM_011521896.2 46 Intron XP_011520198.1
XM_011521899.2 46 Intron XP_011520201.1
XM_017022487.1 46 Intron XP_016877976.1
XM_017022488.1 46 Intron XP_016877977.1

View Full Product Details