Product Details

SNP ID
rs201244472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45135131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCACGTGCTTTCAGACCTGGTGA[A/G]CGTGGAAACTCCCGGCTGCCCCGCC
Phenotype
MIM: 606758 MIM: 612771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUOX1 PubMed Links

Gene Details

Gene
DUOX1
Gene Name
dual oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017434.4 382 Missense Mutation AAC,AGC N112S NP_059130.2
NM_175940.2 382 Missense Mutation AAC,AGC N112S NP_787954.1
XM_011521681.2 382 Missense Mutation AAC,AGC N73S XP_011519983.1
XM_011521682.1 382 Intron XP_011519984.1
Gene
DUOXA1
Gene Name
dual oxidase maturation factor 1
There are no transcripts associated with this gene.

View Full Product Details