Product Details

SNP ID
rs201177341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78593229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGGCAAATATATTAATACCAGT[A/T]CATATTGGAAATGCAAATAAGTGAA
Phenotype
MIM: 118503 MIM: 118505
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CHRNA3 PubMed Links

Gene Details

Gene
CHRNA3
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000743.4 1583 Intron NP_000734.2
NM_001166694.1 1583 Intron NP_001160166.1
XM_006720382.2 1583 Intron XP_006720445.1
Gene
CHRNA5
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000745.3 1583 Silent Mutation GTA,GTT V461V NP_000736.2
NM_001307945.1 1583 UTR 3 NP_001294874.1
XM_005254142.3 1583 UTR 3 XP_005254199.1
XM_017021881.1 1583 UTR 3 XP_016877370.1

View Full Product Details