Product Details

SNP ID
rs200726137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74337994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCACTGCTGGGTTGCTTCCTGG[A/T]TAAAGGGCCAGAAGGTGAAGGAGAT
Phenotype
MIM: 118485
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC33 PubMed Links

Gene Details

Gene
CCDC33
Gene Name
coiled-coil domain containing 33
There are no transcripts associated with this gene.

Gene
CYP11A1
Gene Name
cytochrome P450 family 11 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000781.2 1773 Missense Mutation AAC,ATC N515I NP_000772.2
NM_001099773.1 1773 Missense Mutation AAC,ATC N357I NP_001093243.1

View Full Product Details