Product Details
- SNP ID
-
rs200909165
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:28113094 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTGGATGACGAAGTCTCGGCCCC[A/G]GAAGTCGGCGATGGTCCTGGGCAGC
- Phenotype
-
MIM: 605837
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HERC2
PubMed Links
Gene Details
- Gene
- HERC2
- Gene Name
- HECT and RLD domain containing E3 ubiquitin protein ligase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004667.5 |
7647 |
Missense Mutation |
CGG,TGG |
R4737W |
NP_004658.3 |
XM_005268276.4 |
7647 |
Missense Mutation |
CGG,TGG |
R4699W |
XP_005268333.1 |
XM_006720726.3 |
7647 |
Missense Mutation |
CGG,TGG |
R4732W |
XP_006720789.1 |
XM_006720727.3 |
7647 |
Missense Mutation |
CGG,TGG |
R4651W |
XP_006720790.1 |
XM_011522131.2 |
7647 |
Missense Mutation |
CGG,TGG |
R4576W |
XP_011520433.1 |
XM_011522133.2 |
7647 |
Missense Mutation |
CGG,TGG |
R3652W |
XP_011520435.1 |
XM_017022695.1 |
7647 |
Missense Mutation |
CGG,TGG |
R4699W |
XP_016878184.1 |
XM_017022696.1 |
7647 |
Missense Mutation |
CGG,TGG |
R4699W |
XP_016878185.1 |
XM_017022697.1 |
7647 |
Missense Mutation |
CGG,TGG |
R2459W |
XP_016878186.1 |
XM_017022698.1 |
7647 |
Missense Mutation |
CGG,TGG |
R2459W |
XP_016878187.1 |
View Full Product Details