Product Details

SNP ID
rs199814168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42548902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTGCCAACCCGTGGGACCCGGC[A/G]TCCGCGCCTAACGGCGCTGGGCTAG
Phenotype
MIM: 613429
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAUS2 PubMed Links

Gene Details

Gene
HAUS2
Gene Name
HAUS augmin like complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130447.1 90 Silent Mutation GCA,GCG A10A NP_001123919.1
NM_001323629.1 90 Silent Mutation GCA,GCG A10A NP_001310558.1
NM_001323630.1 90 UTR 5 NP_001310559.1
NM_001323631.1 90 UTR 5 NP_001310560.1
NM_001323632.1 90 UTR 5 NP_001310561.1
NM_018097.2 90 Silent Mutation GCA,GCG A10A NP_060567.1
XM_017022396.1 90 Intron XP_016877885.1
XM_017022397.1 90 Intron XP_016877886.1
Gene
LRRC57
Gene Name
leucine rich repeat containing 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153260.2 90 Intron NP_694992.2
XM_011521423.2 90 Intron XP_011519725.1
XM_011521424.2 90 Intron XP_011519726.1

View Full Product Details