Product Details

SNP ID
rs200659459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45253266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTGCTCTGTCCACAGTGGAGAC[C/T]GGCACAGTGAACCCGGGGCTGGAGC
Phenotype
MIM: 606208
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928414 PubMed Links

Gene Details

Gene
LOC101928414
Gene Name
uncharacterized LOC101928414
There are no transcripts associated with this gene.

Gene
SLC28A2
Gene Name
solute carrier family 28 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004212.3 87 Silent Mutation ACC,ACT T17T NP_004203.2
XM_011522198.2 87 Silent Mutation ACC,ACT T17T XP_011520500.1
XM_011522199.1 87 Silent Mutation ACC,ACT T17T XP_011520501.1
XM_011522200.2 87 Silent Mutation ACC,ACT T17T XP_011520502.1
XM_011522201.2 87 Silent Mutation ACC,ACT T17T XP_011520503.1
XM_011522202.2 87 Intron XP_011520504.1

View Full Product Details