Product Details

SNP ID
rs199752289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65155018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGATTCCCCACTTCGATTAGCAA[A/G]GTCTGCAGCAGCTGCAGCCCTTCTG
Phenotype
MIM: 615611
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLPX PubMed Links

Gene Details

Gene
CLPX
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006660.4 1642 Missense Mutation CTT,TTT L459F NP_006651.2
XM_011521164.2 1642 Intron XP_011519466.1

View Full Product Details