Product Details

SNP ID
rs200327001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19504881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAATAAAAAACCTTCCAACTTAC[A/G]GGGATACAAAATCCTTTTGCATGAG
Phenotype
MIM: 605943
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDE1 PubMed Links

Gene Details

Gene
GDE1
Gene Name
glycerophosphodiester phosphodiesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324066.1 1012 Missense Mutation CCG,CTG P173L NP_001310995.1
NM_001324067.1 1012 Missense Mutation CCG,CTG P252L NP_001310996.1
NM_016641.3 1012 Missense Mutation CCG,CTG P283L NP_057725.1
Gene
TMC5
Gene Name
transmembrane channel like 5
There are no transcripts associated with this gene.

View Full Product Details