Product Details

SNP ID
rs201050211
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70664114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCCCGTGTGGGCCCCATGTAGC[C/T]GGGGGAGTCAGGCACCGTGGGCGTC
Phenotype
MIM: 612081 MIM: 616951
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
IL34 PubMed Links

Gene Details

Gene
IL34
Gene Name
interleukin 34
There are no transcripts associated with this gene.

Gene
MTSS1L
Gene Name
metastasis suppressor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138383.2 2280 Missense Mutation AGC,GGC S603G NP_612392.1
XM_005256236.4 2280 Missense Mutation AGC,GGC S701G XP_005256293.2
XM_011523450.2 2280 Missense Mutation AGC,GGC S647G XP_011521752.1
XM_011523453.2 2280 Missense Mutation AGC,GGC S755G XP_011521755.2
XM_011523454.2 2280 Missense Mutation AGC,GGC S752G XP_011521756.2
XM_011523458.2 2280 Missense Mutation AGC,GGC S474G XP_011521760.1
XM_017023879.1 2280 Missense Mutation AGC,GGC S723G XP_016879368.1
XM_017023880.1 2280 Missense Mutation AGC,GGC S711G XP_016879369.1
XM_017023881.1 2280 Missense Mutation AGC,GGC S704G XP_016879370.1
XM_017023882.1 2280 Intron XP_016879371.1
XM_017023883.1 2280 Intron XP_016879372.1
XM_017023884.1 2280 Intron XP_016879373.1

View Full Product Details