Product Details

SNP ID
rs200920393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:54933842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGACCTGTGCAAAGACTCTCCCT[A/G]TGAATTGAAGAAAGGTATGTCCGAC
Phenotype
MIM: 615624 MIM: 606195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRNDE PubMed Links

Gene Details

Gene
CRNDE
Gene Name
colorectal neoplasia differentially expressed (non-protein coding)
There are no transcripts associated with this gene.

Gene
IRX5
Gene Name
iroquois homeobox 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252197.1 1864 Missense Mutation TAT,TGT Y473C NP_001239126.1
NM_005853.5 1864 Missense Mutation TAT,TGT Y474C NP_005844.4
XM_011522809.1 1864 Missense Mutation TAT,TGT Y404C XP_011521111.1

View Full Product Details