Product Details

SNP ID
rs201147024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:51137402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGATTCCAGAAGCTGGAAGGAT[C/T]CCCACTTCCTGATCTTGCCGCCAAA
Phenotype
MIM: 602218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371253 PubMed Links

Gene Details

Gene
LOC105371253
Gene Name
uncharacterized LOC105371253
There are no transcripts associated with this gene.

Gene
SALL1
Gene Name
spalt like transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127892.1 3716 Missense Mutation AAT,GAT N1132D NP_001121364.1
NM_002968.2 3716 Missense Mutation AAT,GAT N1229D NP_002959.2
XM_006721241.3 3716 Missense Mutation AAT,GAT N1229D XP_006721304.1
XM_011523254.2 3716 Missense Mutation AAT,GAT N1229D XP_011521556.1

View Full Product Details